2'-Deoxyguanosine hydrate ≥99%, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
9-(2-Deoxy-β-D-ribofuranosyl)guanine hydrate, Guanine-2′-deoxyriboside hydrate, 2-Amino-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one hydrate(1:x)
M-Clarity™ quality level = MQ100
854999-100MG
854999-1G
SIAL854999-100MGEA
2840
CZK
SIAL854999-100MG
SIAL854999-1G
2'-Deoxyguanosine hydrate ≥99%, Sigma-Aldrich®
2'-Deoxyguanosine hydrate
Vzorec:
C₁₀H₁₃N₅O₄·xH₂O MW: 267,24 g/mol (anhy) |
MDL Number:
MFCD00150760 CAS číslo: 961-07-9 EINECS: 213-505-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...