Tisk…

Naphthalene black 10B (naphthol blue black, acid black 1), Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Sodium 4-amino-5-hydroxy-3-((4-nitrophenyl)diazenyl)-6-(phenyldiazenyl)naphthalene-2,7-disulphonate, Amido Black 10B

N3393-100G N3393-25G
SIALN3393-100GEA 4320 CZK
SIALN3393-100G SIALN3393-25G
Naphthalene black 10B (naphthol blue black, acid black 1), Sigma-Aldrich®
Naphthalene black 10B (naphthol blue black, acid black 1)
Vzorec: C₂₂H₁₄N₆Na₂O₉S₂
Storage Temperature: Ambient
MDL Number: MFCD00004017
CAS číslo: 1064-48-8
EINECS: 213-903-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR