Tisk…

Naphthalene black 10B (naphthol blue black, acid black 1)

Dodavatel: Thermo Scientific
Varování

Synonyms: Sodium 4-amino-5-hydroxy-3-((4-nitrophenyl)diazenyl)-6-(phenyldiazenyl)naphthalene-2,7-disulphonate, Amido Black 10B

A11374.14 A11374.22
A11374.14EA 1250 CZK
A11374.14 A11374.22
Naphthalene black 10B (naphthol blue black, acid black 1)
Naphthalene black 10B (naphthol blue black, acid black 1)

CAS No.: 1064-48-8 Molecular Formula: C22H14N6Na2O9S2 Formula Weight: 616.49 Odour: Odourless pH: 9.0 Beilstein Reference: 5374263

Vzorec: C₂₂H₁₄N₆Na₂O₉S₂
MW: 616,49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004017
CAS číslo: 1064-48-8
EINECS: 213-903-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR