Tisk…

8-Quinolineboronic acid ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

L19640.03 L19640.MD
L19640.03EA 4330 CZK
L19640.03 L19640.MD
8-Quinolineboronic acid ≥99%
8-Quinolineboronic acid

Purity: 99% CAS No.: 86-58-8 Molecular Formula: C9H8BNO2 Formula Weight: 172.98 Melting Point: 300° Beilstein Reference: 147522 Merck Reference: 14,8069 MDL No.: MFCD01114698

Vzorec: C₉H₈BNO₂
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD01114698
CAS číslo: 86-58-8
Merck Index: 14,8069

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR