MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulphonic acid sodium salt), Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Sodium 2-hydroxy-3-morpholinopropane-1-sulphonate
M-Clarity™ quality level = MQ200
M8767-100G
M8767-500G
SIALM8767-100GEA
10310
CZK
SIALM8767-100G
SIALM8767-500G
MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulphonic acid sodium salt), Sigma-Aldrich®
MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulphonic acid sodium salt)
Vzorec:
C₇H₁₄NNaO₅S MW: 247.2445 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00070007 CAS číslo: 79803-73-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...