MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulphonic acid sodium salt) ≥99%, white powder
Dodavatel: MP Biomedicals
|
Varování
|
Synonyms:
Sodium 2-hydroxy-3-morpholinopropane-1-sulphonate
MOPSO sodium salt is a zwitterionic aminosulfonate buffer that is very similar in structure to MOPS (3-morpholinopropanesulfonic acid), differing by the presence of a hydroxyl group on C-2 of the propane moiety.
MOPSO has been utilized as a buffer component in the analysis of copper by two distinct methods: (1) a flow injection micellar technique of the catalytic reaction of the reaction between 3-methyl-2-benzothiazolinone hydrazone with N-ethyl-N-(2-hydroxy-3-sulfopropyl)-3,5-dimethoxyaniline; 2 (2) an electrospray-ionization quadrapole time-of-flight mass spectroscopy technique to measure the formation of chelating complexes of MOPSO with copper.
Soluble in aqueous solution (5 mg/ml-clear, colorless solution)
Vzorec:
C₇H₁₄NNaO₅S MW: 247.2445 g/mol Bod tání: 270…290 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00070007 CAS číslo: 79803-73-9 |
Výsledky specifikačního testu
Identity Test | Passes |
Purity | ≥99% |
FTIR | Conforms to Standard |
pH (1% aq soln) | 8 - 11 |
Moisture Content | ≤2% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...