Tisk…

1,2-Bis(diphenylphosphino)benzene ≥98%

Dodavatel: Thermo Scientific
Varování

B21106.01 B21106.03
B21106.01EA 1360 CZK
B21106.01 B21106.03
1,2-Bis(diphenylphosphino)benzene ≥98%
1,2-Bis(diphenylphosphino)benzene

Purity: 98% CAS No.: 13991-08-7 Molecular Formula: C26H22P2 Formula Weight: 446.47 Melting Point: 185-189° MDL No.: MFCD00014081

Vzorec: C₃₀H₂₄P₂
MW: 446,46 g/mol
Bod tání: 185…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00014081
CAS číslo: 13991-08-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR