Tisk…

CAPSO sodium salt (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid sodium salt) ≥98%

Dodavatel: Thermo Scientific

Synonyms: Sodium 3-(cyclohexylamino)-2-hydroxypropane-1-sulphonate

B25202.14 B25202.22 B25202.36
B25202.14EA 1320 CZK
B25202.14 B25202.22 B25202.36
CAPSO sodium salt (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid sodium salt) ≥98%
CAPSO sodium salt (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid sodium salt)

Purity: 98% CAS No.: 102601-34-3 Molecular Formula: C9H18NNaO4S Formula Weight: 259.30 MDL No.: MFCD00070063

Vzorec: C₉H₁₈NNaO₄S
MW: 259,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070063
CAS číslo: 102601-34-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR