N-(Hydroxymethyl)-2,2,2-trifluoroacetamide ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2,2,2-Trifluoro-N-(hydroxymethyl)acetamide, N-(Hydroxymethyl)trifluoroacetamide
L00128.14
L00128.06
L00128.14EA
6660
CZK
L00128.14
L00128.06
N-(Hydroxymethyl)-2,2,2-trifluoroacetamide ≥98%
N-(Hydroxymethyl)-2,2,2-trifluoroacetamide
Purity: 98+%
CAS No.: 50667-69-1
Molecular Formula: C3H4F3NO2
Formula Weight: 143.06
Storage Temperature: 2°C to 8°C
Melting Point: 99-103°
Beilstein Reference: 1859914
MDL No.: MFCD00014416
Vzorec:
C₃H₄F₃NO₂ MW: 143,06 g/mol Bod tání: 99…103 °C |
MDL Number:
MFCD00014416 CAS číslo: 50667-69-1 EINECS: 256-698-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...