N-Methyl-bis-trifluoroacetamide, Supelco®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
2,2,2-Trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide, MBTFA
M-Clarity™ quality level = MQ100
M0789-10X1ML
M0789-5ML
M0789-10X1MLEA
10470
CZK
M0789-10X1ML
M0789-5ML
N-Methyl-bis-trifluoroacetamide, Supelco®
N-Methylbistrifluoroacetamide
Vzorec:
C₅H₃F₆NO₂ |
MDL Number:
MFCD00000412 CAS číslo: 685-27-8 EINECS: 211-680-5 UN: 2920 ADR: 8.3,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...