Creatine phosphate disodium salt tetrahydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Sodium creatine phosphate dibasic tetrahydrate, Sodium N-carbamimidoyl-N-methylglycinate dihydrogen phosphate tetrahydrate, Phosphocreatine disodium salt tetrahydrate
27920-1G
27920-5G
SIAL27920-1GEA
2440
CZK
SIAL27920-1G
SIAL27920-5G
Creatine phosphate disodium salt tetrahydrate, Sigma-Aldrich®
Creatine phosphate disodium salt tetrahydrate
Vzorec:
C₄H₈N₃Na₂O₅P·4H₂O MW: 327,14 g/mol (255,07 anhy) Storage Temperature: Freezer |
MDL Number:
MFCD00150192 CAS číslo: 71519-72-7 EINECS: 213-074-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...