Tisk…

Creatine (N-amidinosarcosine) monohydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Creatine Hydrate, 2-(1-Methylguanidino)acetic acid monohydrate, Creatine monohydrate, 2-(1-Methylguanidino)acetic acid hydrate, N-Amidinosarcosine monydrate

M-Clarity™ quality level = MQ200

C3630-100G C3630-1KG
SIALC3630-100GEA 4020 CZK
SIALC3630-100G SIALC3630-1KG
Creatine (N-amidinosarcosine) monohydrate, Sigma-Aldrich®
Creatine (N-amidinosarcosine) monohydrate
Vzorec: C₄H₉N₃O₂·1H₂O
MW: 149,15 g/mol (131,13 anhy)
MDL Number: MFCD00071582
CAS číslo: 6020-87-7
EINECS: 200-306-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR