Creatine (N-amidinosarcosine) monohydrate ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Creatine Hydrate, 2-(1-Methylguanidino)acetic acid monohydrate, Creatine monohydrate, 2-(1-Methylguanidino)acetic acid hydrate, N-Amidinosarcosine monydrate
B25009.0E
B25009.22
B25009.36
B25009.0EEA
24300
CZK
B25009.0E
B25009.22
B25009.36
Creatine (N-amidinosarcosine) monohydrate ≥99%
Creatine (N-amidinosarcosine) monohydrate
Purity: 99%
CAS No.: 6020-87-7
Molecular Formula: C4H9N3O2.H2O
Formula Weight: 149.15 (131.13anhy)
Odour: Weak
pH: 6.9
Melting Point: ca 292° dec.
Beilstein Reference: 907175
Merck Reference: 14,2568
MDL No.: MFCD00071582
Vzorec:
C₄H₉N₃O₂·1H₂O MW: 149,15 g/mol (131,13 anhy) Bod tání: ∼ 292 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00071582 CAS číslo: 6020-87-7 EINECS: 200-306-6 Merck Index: 14,2568 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...