Tisk…
L(+)-Glutamic acid hydrochloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: (S)-2-Aminopentanedioic acid hydrochloride, 2-Aminoglutaric acid hydrochloride
M-Clarity™ quality level = MQ200
G2128-100G G2128-1KG G2128-500G
SIALG2128-100GEA 940 CZK
SIALG2128-100G SIALG2128-1KG SIALG2128-500G
L(+)-Glutamic acid hydrochloride, Sigma-Aldrich®
L(+)-Glutamic acid hydrochloride
Vzorec: C₅H₁₀ClNO₄
MW: 183.59 g/mol
MDL Number: MFCD00012619
CAS číslo: 138-15-8
EINECS: 205-315-9