N-Decanoyl-N-methyl-D-glucamine (MEGA-10), Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
D6277-1G
D6277-5G
SIALD6277-1GEA
6150
CZK
SIALD6277-1G
SIALD6277-5G
N-Decanoyl-N-methyl-D-glucamine (MEGA-10), Sigma-Aldrich®
N-Decanoyl-N-methyl-D-glucamine (MEGA-10)
Vzorec:
C₁₇H₃₅NO₆ MW: 349.46 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00036801 CAS číslo: 85261-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...