Tisk…

o-Cresolphthalein complexone tetrasodium salt, pure

Dodavatel: Thermo Scientific

405750050 405750250
ACRO405750050EA 1860 CZK
ACRO405750050 ACRO405750250
o-Cresolphthalein complexone tetrasodium salt, pure
o-Cresolphthalein complexone tetrasodium salt
Vzorec: C₃₂H₂₈N₂Na₄O₁₂
Storage Temperature: Ambient
MDL Number: MFCD00066371
CAS číslo: 62698-54-8

Objednat

Výsledky specifikačního testu

Appearance Purple Powder or crystals
Infrared spectrum Conforms
Suitability (for use as reagent for Ca) Passes test
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR