o-Cresolphthalein complexone, pure
Dodavatel: Thermo Scientific
182440050
182440250
182441000
ACRO182440050EA
4200
CZK
ACRO182440050
ACRO182440250
ACRO182441000
o-Cresolphthalein complexone, pure
o-Cresolphthalein complexone
Vzorec:
C₃₂H₃₂N₂O₁₂ Bod tání: 181 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00005911 CAS číslo: 2411-89-4 EINECS: 219-318-8 |
Výsledky specifikačního testu
Appearance | Off-white to pink-beige Powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Loss on drying | ≤10 % (1 g, 105°C) |
Sulfated ash | ≤0.5 % |
UV Lambda max | 573 to 579 nm (in 0.1N NaOH) |
Suitability | (as complexom. indicator) Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...