Tisk…

Erythromycin, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: (-)-Erythromycin

M-Clarity™ quality level = MQ200

E6376-100G E6376-25G
SIALE6376-100GEA 19580 CZK
SIALE6376-100G SIALE6376-25G
Erythromycin, Sigma-Aldrich®
Erythromycin
Vzorec: C₃₇H₆₇NO₁₃
Storage Temperature: Ambient
MDL Number: MFCD00084654
CAS číslo: 114-07-8
EINECS: 204-040-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR