Tisk…

Brilliant green, pure, high purity biological stain

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate

229601000 229605000
ACRO229601000EA 1820 CZK
ACRO229601000 ACRO229605000
Brilliant green, pure, high purity biological stain
Brilliant green
Vzorec: C₂₇H₃₄N₂O₄S
Bod tání: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00011880
CAS číslo: 633-03-4
EINECS: 211-190-1

Objednat

Výsledky specifikačního testu

Appearance Green to green-yellow Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤5 % (105°C)
UV Lambda max 625 to 631 nm (in 50% ethanol)
Absorption Ratio P-15/P+15 0.94 to 1.14
Specific absorbance A (1%/1cm) ≥1900 (at 625 to 631 nm)
Visual transition interval from pH 0.0 (yellow) to pH 2.6 (green)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR