Tisk…

Methyl green zinc chloride salt, pure, certified

Dodavatel: Thermo Fisher Scientific
Varování

414380250 414381000
ACRO414380250EA 3760 CZK
ACRO414380250 ACRO414381000
Methyl green zinc chloride salt, pure, certified
Methyl green zinc chloride salt
Vzorec: C₂₇H₃₅BrClN₃·xZnCl₂
Storage Temperature: Ambient
MDL Number: MFCD00151094
CAS číslo: 7114-03-6
EINECS: 230-415-4

Objednat

Výsledky specifikačního testu

Appearance Dark green to dark brown-red Powder
Infrared spectrum Conforms
Dye content ≥85 %
UV Lambda max 630 to 635 nm (in water)
Absorption Ratio P-15/P+15 0.86 to 1.11
Visual transition interval from pH 0.2 (yellow) to pH 1.8 (blue)
BSC Certification Passes

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR