Tisk…

(±)-Tetrahydro-2-furoic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Tetrahydrofuran-2-carboxylic acid, 2-Tetrahydrofuran Carboxylic Acid

M-Clarity™ quality level = MQ100

341517-25G 341517-5G
SIAL341517-25GEA 2670 CZK
SIAL341517-25G SIAL341517-5G
(±)-Tetrahydro-2-furoic acid, Sigma-Aldrich®
(±)-Tetrahydro-2-furoic acid
Vzorec: C₅H₈O₃
MW: 116,12 g/mol
MDL Number: MFCD00022473
CAS číslo: 16874-33-2
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR