(±)-Tetrahydro-2-furoic acid ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Tetrahydrofuran-2-carboxylic acid, 2-Tetrahydrofuran Carboxylic Acid
B22927.14
B22927.22
B22927.14EA
1520
CZK
B22927.14
B22927.22
(±)-Tetrahydro-2-furoic acid ≥97%
(±)-Tetrahydro-2-furoic acid
Purity: 97+%
CAS No.: 16874-33-2
Molecular Formula: C5H8O3
Formula Weight: 116.12
Flash Point: 139°(282°F)
Refractive Index: 1.4600
Odour: Pungent
Boiling Point: 244°
Beilstein Reference: 109671
UN No.: 3265
MDL No.: MFCD00022473
Vzorec:
C₅H₈O₃ MW: 116,12 g/mol Bod varu: 244 °C Hustota: 1,213 Bod vzplanutí: 139 °C (282 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00022473 CAS číslo: 16874-33-2 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...