Tisk…

Carbenicillin disodium salt, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

C9231-10G C9231-1G C9231-5G
SIALC9231-10GEA 0 CZK
SIALC9231-10G SIALC9231-1G SIALC9231-5G
Carbenicillin disodium salt, Sigma-Aldrich®
Carbenicillin disodium salt
Vzorec: C₁₇H₁₆N₂O₆S·2Na
Storage Temperature: Refrigerator
MDL Number: MFCD00077683
CAS číslo: 4800-94-6
EINECS: 225-360-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR