Carbenicillin disodium salt, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
NSC 111071, α-Carboxybenzylpenicillin disodium salt, Sodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na
M-Clarity™ quality level = MQ200
C1389-10G
C1389-1G
C1389-250MG
C1389-5G
SIALC1389-10GEA
0
CZK
SIALC1389-10G
SIALC1389-1G
SIALC1389-250MG
SIALC1389-5G
Carbenicillin disodium salt, Sigma-Aldrich®
Carbenicillin disodium salt
Vzorec:
C₁₇H₁₆N₂O₆S·2Na MW: 422,363 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00077683 CAS číslo: 4800-94-6 EINECS: 225-360-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...