Tisk…

2,6-Diaminopurine 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 9H-Purine-2,6-diamine, 7H-Purine-2,6-diamine

A15649.03 A15649.06
A15649.03EA 985 CZK
A15649.03 A15649.06
2,6-Diaminopurine 98%
2,6-Diaminopurine

Purity: 98% CAS No.: 1904-98-9 Molecular Formula: C5H6N6 Formula Weight: 150.14 Melting Point: ca 302° Beilstein Reference: 9624 Merck Reference: 14,2985 MDL No.: MFCD00213668

Vzorec: C₅H₆N₆
MW: 150,14 g/mol
Bod tání: ∼ 302 °C
Storage Temperature: Ambient
MDL Number: MFCD00213668
CAS číslo: 1904-98-9
EINECS: 217-605-2
Merck Index: 14,2985

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR