Tisk…

Diphenylphosphinic acid ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

A11563.06 A11563.14 A11563.22
A11563.06EA 1640 CZK
A11563.06 A11563.14 A11563.22
Diphenylphosphinic acid ≥99%
Diphenylphosphinic acid

Purity: 99% CAS No.: 1707-03-5 Molecular Formula: C12H11O2P Formula Weight: 218.19 Melting Point: 193-197° Beilstein Reference: 2804567

Vzorec: C₁₂H₁₁O₂P
Bod tání: 193…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00002132
CAS číslo: 1707-03-5
EINECS: 216-948-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR