Diphenylphosphinic chloride 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Diphenylphosphinyl chloride
212960100
212960500
ACRO212960100EA
2160
CZK
ACRO212960100
ACRO212960500
Diphenylphosphinic chloride 98%
Diphenylphosphinic chloride
Vzorec:
C₁₂H₁₀ClOP Bod varu: 222 °C (16 mmHg) Bod vzplanutí: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002077 CAS číslo: 1499-21-4 EINECS: 216-107-2 UN: 2920 ADR: 8,II |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6080 to 1.6120 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...