Tricyclohexylphosphonium tetrafluoroborate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
631493-1G
631493-5G
SIAL631493-1GEA
1650
CZK
SIAL631493-1G
SIAL631493-5G
Tricyclohexylphosphonium tetrafluoroborate, Sigma-Aldrich®
Tricyclohexylphosphonium tetrafluoroborate
Vzorec:
C₁₈H₃₄BF₄P MW: 368,24 g/mol |
MDL Number:
MFCD03840580 CAS číslo: 58656-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...