Tricyclohexylphosphonium tetrafluoroborate ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
H27428.03
H27428.14
H27428.03EA
1300
CZK
H27428.03
H27428.14
Tricyclohexylphosphonium tetrafluoroborate ≥99%
Tricyclohexylphosphonium tetrafluoroborate
Purity: 99%
CAS No.: 58656-04-5
Molecular Formula: C18H34P·BF4
Formula Weight: 368.25
Melting Point: 166-170°
MDL No.: MFCD03840580
Vzorec:
C₁₈H₃₄BF₄P MW: 368,24 g/mol Bod tání: 166…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840580 CAS číslo: 58656-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...