(+)-Dibenzoyl-D-tartaric acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(2S,3S)-(+)-Di-O-benzoyltartaric acid, (2S,3S)-2,3-Bis(benzoyloxy)succinic acid
M-Clarity™ quality level = MQ100
163449-10G
163449-50G
SIAL163449-10GEA
719
CZK
SIAL163449-10G
SIAL163449-50G
(+)-Dibenzoyl-D-tartaric acid, Sigma-Aldrich®
(+)-Dibenzoyl-D-tartaric acid
Vzorec:
C₁₈H₁₄O₈ MW: 358,31 g/mol |
MDL Number:
MFCD00063222 CAS číslo: 17026-42-5 EINECS: 241-097-1 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...