(+)-Dibenzoyl-D-tartaric acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(2S,3S)-(+)-Di-O-benzoyltartaric acid, (2S,3S)-2,3-Bis(benzoyloxy)succinic acid
33610-250G-F
33610-50G-F
SIAL33610-250G-FEA
12350
CZK
SIAL33610-250G-F
SIAL33610-50G-F
(+)-Dibenzoyl-D-tartaric acid, Sigma-Aldrich®
(+)-Dibenzoyl-D-tartaric acid
Vzorec:
C₁₈H₁₄O₈ MW: 358,31 g/mol |
MDL Number:
MFCD00063222 CAS číslo: 17026-42-5 EINECS: 241-097-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...