Tisk…

(+)-Di-p-toluoyl-D-tartaric acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Di-p-toluoyl-D-tartaric acid, (2S,3S)-(+)-Di-O-4-Toluoyl-D-Tartaric Acid, Di-p-toluoyl-D-tartaric Acid

302813-100G 302813-25G
SIAL302813-100GEA 7900 CZK
SIAL302813-100G SIAL302813-25G
(+)-Di-p-toluoyl-D-tartaric acid, Sigma-Aldrich®
(+)-Di-p-toluoyl-D-tartaric acid
Vzorec: C₂₀H₁₈O₈
MW: 386,36 g/mol
MDL Number: MFCD00008552
CAS číslo: 32634-68-7
EINECS: 251-132-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR