Tisk…

(-)-Dibenzoyl-L-tartaric acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (2R,3R)-2,3-Bis(benzoyloxy)succinic acid

M-Clarity™ quality level = MQ100

345849-100G 345849-25G
SIAL345849-100GEA 3740 CZK
SIAL345849-100G SIAL345849-25G
(-)-Dibenzoyl-L-tartaric acid, Sigma-Aldrich®
(-)-Dibenzoyl-L-tartaric acid
Vzorec: C₁₈H₁₄O₈
MW: 358,31 g/mol
MDL Number: MFCD00003074
CAS číslo: 2743-38-6
EINECS: 220-374-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR