Alizarin red S (sodium salt), pure, certified
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulphonate, C.I. 58005, Alizarin sulphonic acid disodium salt, Alizarin Red S sodium
400480250
400481000
ACRO400480250EA
4380
CZK
ACRO400480250
ACRO400481000
Alizarin red S (sodium salt), pure, certified
Alizarin Red S (sodium salt)
Vzorec:
C₁₄H₇NaO₇S MW: 342,26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013049 CAS číslo: 130-22-3 EINECS: 204-981-8 |
Výsledky specifikačního testu
Appearance | Yellow to orange to brown Powder |
Infrared spectrum | Conforms |
UV Lambda max | 590 to 599 nm |
Visual transition interval | from pH 9.4 (orange-brown) to pH 12.0 (violet) |
BSC Certification | Passes |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...