Tisk…

Spermine tetrahydrochloride, Sigma-Aldrich®

Dodavatel: Merck

85605-1G 85605-25G 85605-5G
SIAL85605-1GEA 4320 CZK
SIAL85605-1G SIAL85605-25G SIAL85605-5G
Spermine tetrahydrochloride, Sigma-Aldrich®
Spermine tetrahydrochloride
Vzorec: C₁₀H₃₀Cl₄N₄
MDL Number: MFCD00012914
CAS číslo: 306-67-2
EINECS: 206-189-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR