Tisk…

Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

S3256-1G S3256-25G S3256-5G
SIALS3256-1GEA 3690 CZK
SIALS3256-1G SIALS3256-25G SIALS3256-5G
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine), Sigma-Aldrich®
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine)
Vzorec: C₁₀H₂₆N₄
MDL Number: MFCD00008215
CAS číslo: 71-44-3
EINECS: 200-754-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR