N-Acetylglycine(Ac-Gly-OH), Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Ac-Gly-OH, N-Acetylglycine, Acetamidoacetic acid, Aceturic acid
M-Clarity™ quality level = MQ200
A16300-100G
A16300-500G
A16300-5G
SIALA16300-100GEA
919
CZK
SIALA16300-100G
SIALA16300-500G
SIALA16300-5G
N-Acetylglycine(Ac-Gly-OH), Sigma-Aldrich®
N-Acetylglycine (Ac-Gly-OH)
Vzorec:
C₄H₇NO₃ MW: 117,1 g/mol |
MDL Number:
MFCD00004275 CAS číslo: 543-24-8 EINECS: 208-839-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...