7-Phenyl-1-heptanol ≥97%
Dodavatel: Thermo Fisher Scientific
L12533.03
L12533.06
L12533.03EA
2860
CZK
L12533.03
L12533.06
7-Phenyl-1-heptanol ≥97%
7-Phenyl-1-heptanol
Purity: 97%
CAS No.: 3208-25-1
Molecular Formula: C6H5(CH2)7OH
Formula Weight: 192.30
Refractive Index: 1.5080
Boiling Point: 115-117°/0.5mm
MDL No.: MFCD00037315
Vzorec:
C₁₃H₂₀O Bod varu: 115…117 °C (0,5 mmHg) Hustota: 0,962 Storage Temperature: Ambient |
MDL Number:
MFCD00037315 CAS číslo: 3208-25-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...