Tisk…

ß-Nicotinamide adenine dinucleotide phosphate, oxidised form (NADP, oxidised form) hydrate, Sigma-Aldrich®

Dodavatel: Merck

N5755-100MG N5755-500MG N5755-1G
SIALN5755-100MGEA 8840 CZK
SIALN5755-100MG SIALN5755-500MG SIALN5755-1G
ß-Nicotinamide adenine dinucleotide phosphate, oxidised form (NADP, oxidised form) hydrate, Sigma-Aldrich®
ß-Nicotinamide adenine dinucleotide phosphate, oxidised form (NADP, oxidised form) hydrate
Vzorec: C₂₁H₂₈N₇O₁₇P₃·H₂O
CAS číslo: 53-59-8
EINECS: 200-178-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR