β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) hydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
beta-NADH disodium salt hydrate, beta-Nicotinamide adenine dinucleotide disodium salt hydrate, β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form) hydrate
N8129-100MG
N8129-500MG
N8129-50MG
N8129-1G
N8129-5G
SIALN8129-100MGEA
2620
CZK
SIALN8129-100MG
SIALN8129-500MG
SIALN8129-50MG
SIALN8129-1G
SIALN8129-5G
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) hydrate, Sigma-Aldrich®
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) hydrate
Vzorec:
C₂₁H₂₇N₇Na₂O₁₄P₂·H₂O MW: 727.43 g/mol |
MDL Number:
MFCD03791273 CAS číslo: 606-68-8 EINECS: 210-123-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...