β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Cozymase, beta-DPN, Coenzyme I, beta-Nicotinamide Adenine dinucleotide hydrate, oxidized form, beta-Diphosphopyridine Nucleotide, beta-NAD
N6522-250MG
N6522-1G
N6522-5G
SIALN6522-250MGEA
3600
CZK
SIALN6522-250MG
SIALN6522-1G
SIALN6522-5G
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate, Sigma-Aldrich®
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate
Vzorec:
C₂₁H₂₇N₇O₁₄P₂·H₂O |
MDL Number:
MFCD00036253 CAS číslo: 53-84-9 EINECS: 200-184-4 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...