Tisk…
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Cozymase, beta-DPN, Coenzyme I, beta-Nicotinamide Adenine dinucleotide hydrate, oxidized form, beta-Diphosphopyridine Nucleotide, beta-NAD
43410-250MG 43410-1G 43410-5G
SIAL43410-250MGEA 1030 CZK
SIAL43410-250MG SIAL43410-1G SIAL43410-5G
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate, Sigma-Aldrich®
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate
Vzorec: C₂₁H₂₇N₇O₁₄P₂·H₂O
MDL Number: MFCD00036253
CAS číslo: 53-84-9
EINECS: 200-184-4