m-Cresol purple, pure indicator grade
Dodavatel: Thermo Fisher Scientific
Synonyms:
2',2''-Dimethylphenolsulphonphthalein, m-Cresolsulphonphthalein, MCP, 3,3-Bis(4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
228200050
228200100
228200500
ACRO228200050EA
1670
CZK
ACRO228200050
ACRO228200100
ACRO228200500
m-Cresol purple, pure indicator grade
m-Cresol purple
Vzorec:
C₂₁H₁₈O₅S MW: 382.43 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005871 CAS číslo: 2303-01-7 EINECS: 218-960-6 |
Výsledky specifikačního testu
Appearance | Dark green Crystalline powder |
Infrared spectrum | Conforms |
Solubility | (0.1 % in 50 % ethanol) Clear |
Specific extinction E (1%/1cm) | ≥900 (578 nm, in buffer) |
Visual transition interval | from pH 7.4 (yellow) to pH 9.0 (purple) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...