Tisk…

Octyl-α-D-glucopyranoside, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl α-glucoside, Octyl α-D-glucopyranoside

M-Clarity™ quality level = MQ200

O0630-100MG O0630-1G
SIALO0630-100MGEA 10590 CZK
SIALO0630-100MG SIALO0630-1G
Octyl-α-D-glucopyranoside, Sigma-Aldrich®
Octyl-α-D-glucopyranoside
Vzorec: C₁₄H₂₈O₆
MW: 292,37 g/mol
MDL Number: MFCD00070008
CAS číslo: 29781-80-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR