Tisk…

α-D-Glucose, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

158968-25G 158968-500G
SIAL158968-25GEA 582 CZK
SIAL158968-25G SIAL158968-500G
α-D-Glucose, Sigma-Aldrich®
α-D-Glucose
Vzorec: C₆H₁₂O₆
MDL Number: MFCD00063774
CAS číslo: 492-62-6
EINECS: 207-757-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR