6-Deoxy-D-glucose, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
D9761-100MG
D9761-250MG
SIALD9761-100MGEA
11240
CZK
SIALD9761-100MG
SIALD9761-250MG
6-Deoxy-D-glucose, Sigma-Aldrich®
6-Deoxy-D-glucose
Vzorec:
C₆H₁₂O₅ |
MDL Number:
MFCD00061627 CAS číslo: 7658-08-4 EINECS: 231-622-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...