N-Acetyl-β-D-glucosamine 98%
Dodavatel: Thermo Scientific
Synonyms:
2-Acetamido-2-deoxy-D-glucopyranose, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc, N-Acetyl-D-glucosamine
270940100
270941000
270945000
ACRO270940100EA
997
CZK
ACRO270940100
ACRO270941000
ACRO270945000
N-Acetyl-β-D-glucosamine 98%
N-Acetyl-β-D-glucosamine
Vzorec:
C₈H₁₅NO₆ MW: 221,21 g/mol Bod tání: 211 °C (rozkl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00136044 CAS číslo: 7512-17-6 EINECS: 231-368-2 |
Výsledky specifikačního testu
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % (105°C, 4 hrs) |
HPLC | ≥97.5 % (ELSD) |
Specific optical rotation | +39° to +45° (20°C, 589 nm) (c=2, H2O, after 2 hrs) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...