Tisk…

3-Fluoro-4-hydroxybenzaldehyde ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

L19325.03 L19325.06
L19325.03EA 2790 CZK
L19325.03 L19325.06
3-Fluoro-4-hydroxybenzaldehyde ≥98%
3-Fluoro-4-hydroxybenzaldehyde

Purity: 98% CAS No.: 405-05-0 Molecular Formula: C7H5FO2 Formula Weight: 140.11 Melting Point: 121-124° Beilstein Reference: 3234259

Vzorec: C₇H₅FO₂
Bod tání: 121…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00016626
CAS číslo: 405-05-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR