Tisk…

Tiglic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (E)-2-Methylbut-2-enoic acid, trans-2,3-Dimethylacrylic acid

M-Clarity™ quality level = MQ300

W359904-100G-K W359904-1KG-K W359904-5KG-K
SIALW359904-100G-KEA 3320 CZK
SIALW359904-100G-K SIALW359904-1KG-K SIALW359904-5KG-K
Tiglic acid, Sigma-Aldrich®
Tiglic acid
Vzorec: C₅H₈O₂
MW: 100,12 g/mol
MDL Number: MFCD00066864
CAS číslo: 80-59-1
EINECS: 201-295-0
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR