Tisk…

Tiglic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: (E)-2-Methylbut-2-enoic acid, trans-2,3-Dimethylacrylic acid

A13905.14 A13905.22 A13905.36
A13905.14EA 903 CZK
A13905.14 A13905.22 A13905.36
Tiglic acid ≥98%
Tiglic acid

Purity: 98+% CAS No.: 80-59-1 Molecular Formula: C5H8O2 Formula Weight: 100.12 Flash Point: 101°(213°F) Melting Point: 62-66° Boiling Point: 198-199° Beilstein Reference: 1236500 Merck Reference: 14,9433 UN No.: 3261 MDL No.: MFCD00066864

Vzorec: C₅H₈O₂
Bod varu: 198…199 °C
Bod tání: 62…66 °C
Hustota: 0,969
Bod vzplanutí: 101 °C (213 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066864
CAS číslo: 80-59-1
EINECS: 201-295-0
UN: 3261
ADR: 8,III
Merck Index: 14,9433

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR